APO456796247 Apollo Scientific Ltd

2,4-Bis(trifluoromethyl)aniline

  • MDL number MFCD00236209
  • Cas Number : 367-71-5
  • Molecular Weight : 229.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,4-bis(trifluoromethyl)phenylamine
Inchi Code 1S/C8H5F6N/c9-7(10,11)4-1-2-6(15)5(3-4)8(12,13)14/h1-3H,15H2
Inchi Key UIWVOUBVGBJRNP-UHFFFAOYSA-N

Description

Flash Point 50/0.4mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-bistrifluoromethylaniline

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/24-bistrifluoromethylaniline
pictograms GHS07;GHS08
hstatements H335;H412
Availability details for
Quantity
  • Availability
  • Add to Cart