APO456796256 Apollo Scientific Ltd

2-Chloro-1,1,2-trifluoroethyl methyl ether

  • MDL number MFCD00039335
  • Cas Number : 425-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -92
Synonym 2-Chloro-1,1,2-trifluoro-1-methoxyethane, 1-Chloro-1,2,2-trifluoro-3-oxabutane
Purity 97
Density 1.363
Flash Point 66
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 69-71.5
Product Link http://www.apolloscientific.co.uk/display_item.php?id=980

Safety Information

signalword Danger
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=980
pictograms GHS06
hstatements H311;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart