APO456796260 Apollo Scientific Ltd

2-Chloro-5-(trifluoromethyl)phenyl isocyanate

  • MDL number MFCD00037029
  • Cas Number : 50528-86-4
  • Molecular Weight : 221.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 92
Iupac Name 1-chloro-2-isocyanato-4-(trifluoromethyl)benzene
density 1.48
Inchi Code 1S/C8H3ClF3NO/c9-6-2-1-5(8(10,11)12)3-7(6)13-4-14/h1-3H
Inchi Key WEPYOPYMWSHRIW-UHFFFAOYSA-N

Description

Melting Point 92
Synonym 4-Chloro-3-isocyanatobenzotrifluoride, 1-Chloro-2-isocyanato-4-(trifluoromethyl)benzene
Purity 98%
Density 1.48
Flash Point 63-64/5mm
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-5-trifluoromethylphenyl-isocyanate

Safety Information

signalword Danger
pcodes P271;P280;P260
MSDS https://store.apolloscientific.co.uk/product/2-chloro-5-trifluoromethylphenyl-isocyanate
pictograms GHS06;GHS08
hstatements H301+H311+H331;H317;H319
Availability details for
Quantity
  • Availability
  • Add to Cart