APO456796264 Apollo Scientific Ltd

3-Chloro-2-thio-5-(trifluoromethyl)pyridine

  • MDL number MFCD00153104
  • Cas Number : 76041-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-128
Synonym 3-Chloro-2-sulphanyl-5-(trifluoromethyl)pyridine, 3-Chloro-2-mercapto-5-(trifluoromethyl)pyridine, 3-Chloro-5-(trifluoromethyl)pyridine-2-thiol
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1016

Safety Information

signalword Warning
pcodes P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1016
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart