APO456796305 Apollo Scientific Ltd

2,4-Dibromo-6-fluoroaniline

  • MDL number MFCD00042230
  • Cas Number : 141474-37-5
  • Molecular Weight : 268.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62-64
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-dibromo-6-fluoroaniline

Safety Information

signalword Warning
pcodes P260;P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/24-dibromo-6-fluoroaniline
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart