APO456796308 Apollo Scientific Ltd

2,6-Dibromo-4-fluoroiodobenzene

  • MDL number MFCD00042232
  • Cas Number : 62720-29-0
  • Molecular Weight : 379.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128-130
Synonym 1,3-Dibromo-5-fluoro-2-iodobenzene
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-dibromo-4-fluoroiodobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/26-dibromo-4-fluoroiodobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart