APO456796351 Apollo Scientific Ltd

2-(Phenylthio)-5-(trifluoromethyl)benzoic acid

  • MDL number MFCD00069137
  • Cas Number : 52548-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153-154
Synonym 2-Carboxy-4-(trifluoromethyl)diphenyl sulphide, 2-(Phenylsulphanyl)-5-(trifluoromethyl)benzoic acid
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10300

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10300
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart