APO456796357 Apollo Scientific Ltd

2,3-Difluoro-4-nitroanisole

  • MDL number MFCD07995055
  • Cas Number : 66684-59-1
  • Molecular Weight : 189.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96-99
Synonym 2,3-Difluoro-4-methoxynitrobenzene, 2,3-Difluoro-4-nitrophenyl methyl ether
Purity 98
Flash Point >110
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/23-difluoro-4-nitroanisole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/23-difluoro-4-nitroanisole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart