APO456796385 Apollo Scientific Ltd

2-Bromo-4-fluoro-6-nitrophenol

  • MDL number MFCD03094182
  • Cas Number : 320-75-2
  • Molecular Weight : 236

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 98
bp 67
Iupac Name 2-bromo-4-fluoro-6-nitrophenol
Inchi Code 1S/C6H3BrFNO3/c7-4-1-3(8)2-5(6(4)10)9(11)12/h1-2,10H
Inchi Key DFHGOCSWFGXYSQ-UHFFFAOYSA-N

Description

Melting Point 98
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 67
Product Link https://store.apolloscientific.co.uk/product/2-bromo-4-fluoro-6-nitrophenol

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-4-fluoro-6-nitrophenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart