APO456796410 Apollo Scientific Ltd

Silver perfluorooctanoate

  • MDL number MFCD00040393
  • Cas Number : 335-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 252-261
Synonym Silver pentadecafluorooctanoate, Silver pentadecafluorocaprylate
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5787

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5787
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart