APO456796443 Apollo Scientific Ltd

Ethyl 5-chloro-2-fluorobenzoate

  • MDL number MFCD06205125
  • Cas Number : 773139-56-3
  • Molecular Weight : 202.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-chloro-2-fluorobenzoate
Inchi Code 1S/C9H8ClFO2/c1-2-13-9(12)7-5-6(10)3-4-8(7)11/h3-5H,2H2,1H3
Inchi Key DQIQQERKWZQXCN-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-5-chloro-2-fluorobenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-5-chloro-2-fluorobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart