APO456796460 Apollo Scientific Ltd

Cyclopropyl(4-fluorophenyl)methanone

  • MDL number MFCD00013728
  • Cas Number : 772-31-6
  • Molecular Weight : 164.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Cyclopropyl 4-fluorophenyl ketone, (4-Fluorobenzoyl)cyclopropane
Purity 95
Density 1.14
Flash Point 100
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 119-120/13mm
Product Link https://store.apolloscientific.co.uk/product/cyclopropyl4-fluorophenylmethanone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/cyclopropyl4-fluorophenylmethanone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart