APO456796484 Apollo Scientific Ltd

3',5'-Bis(trifluoromethyl)propiophenone

  • MDL number MFCD00009911
  • Cas Number : 85068-34-4
  • Molecular Weight : 270.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.379
Flash Point 76
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 72/7mm
Product Link https://store.apolloscientific.co.uk/product/35-bistrifluoromethylpropiophenone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/35-bistrifluoromethylpropiophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart