APO456796521 Apollo Scientific Ltd

Ethyl 4-(trifluoromethyl)benzoate

  • MDL number MFCD00013563
  • Cas Number : 583-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 78-80/5mm
Product Link https://store.apolloscientific.co.uk/product/ethyl-4-trifluoromethylbenzoate-97

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-4-trifluoromethylbenzoate-97
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart