APO456796546 Apollo Scientific Ltd

2-[(2-Aminoethyl)amino]-3-chloro-5-(trifluoromethyl)pyridine

  • MDL number MFCD00099696
  • Cas Number : 219478-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]ethane-1,2-diamine, 6-[(2-Aminoethyl)amino]-5-chloro-alpha,alpha,alpha-trifluoro-3-picoline
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 96/0.3mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=7334

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=7334
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart