APO456796555 Apollo Scientific Ltd

4-(Bromomethyl)-1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD09879947
  • Cas Number : 912569-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75.5-77.5
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=42253

Safety Information

signalword Danger
pcodes P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=42253
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart