APO456796578 Apollo Scientific Ltd

N1-Methyl-4-(trifluoromethyl)benzene-1,2-diamine

  • MDL number MFCD01571385
  • Cas Number : 35203-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-55
Synonym 3-Amino-4-(methylamino)benzotrifluoride, 2-Amino-1-(methylamino)-4-(trifluoromethyl)benzene
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/n1-methyl-4-trifluoromethylbenzene-12-diamine

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/n1-methyl-4-trifluoromethylbenzene-12-diamine
pictograms GHS06
hstatements H301+H311+H331;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart