APO456796580 Apollo Scientific Ltd

(2R)-(+)-3,3,3-Trifluoro-1,2-propenoxide

  • MDL number MFCD08276876
  • Cas Number : 143142-90-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -80
Synonym (2R)-2-(Trifluoromethyl)oxirane, (2R)-1,2-Epoxy-3,3,3-trifluoropropane
Density 1.294
Flash Point -26
Storage temp. Store at 2-8
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 25-32
Product Link https://store.apolloscientific.co.uk/product/2r-333-trifluoro-12-propenoxide

Safety Information

signalword TBC
pcodes TBC
MSDS https://store.apolloscientific.co.uk/product/2r-333-trifluoro-12-propenoxide
pictograms TBC
hstatements TBC
Availability details for
Quantity
  • Availability
  • Add to Cart