APO456796631 Apollo Scientific Ltd

Sodium bromo(difluoro)acetate

  • MDL number MFCD04038355
  • Cas Number : 84349-27-9
  • Molecular Weight : 196.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Bromo(difluoro)acetic acid, sodium salt, Sodium 2-bromo-2,2-difluoroethanoate
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/sodium-bromodifluoroacetate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/sodium-bromodifluoroacetate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart