APO456796644 Apollo Scientific Ltd

2-(Trifluoromethyl)-3-ethoxydodecafluorohexane

  • MDL number MFCD04038367
  • Cas Number : 297730-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -100
Synonym 3-Ethoxyperfluoro(2-methylhexane)
Purity 99
Density 1.61
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 130
Product Link https://store.apolloscientific.co.uk/product/2-trifluoromethyl-3-ethoxydodecafluorohexane-99

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-trifluoromethyl-3-ethoxydodecafluorohexane-99
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart