APO456796790 Apollo Scientific Ltd

3-Chloro-2-fluoroprop-1-ene

  • MDL number MFCD01696350
  • Cas Number : 6186-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Fluoroallyl chloride, 2-Fluoroprop-2-en-1-yl chloride
Purity 95
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 55
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2736

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2736
pictograms GHS02;GHS07
hstatements H226;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart