APO456796817 Apollo Scientific Ltd

Octafluoroadipoyl chloride

  • MDL number MFCD00054726
  • Cas Number : 336-06-1
  • Molecular Weight : 326.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Perfluoroadipoyl chloride
Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 42-45/2mm
Product Link https://store.apolloscientific.co.uk/product/octafluoroadipoyl-chloride

Safety Information

signalword Danger
pcodes P232;P260;P280
MSDS https://store.apolloscientific.co.uk/product/octafluoroadipoyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart