APO456796823 Apollo Scientific Ltd

3-Cyano-5-fluorobenzoic acid

  • MDL number MFCD05663717
  • Cas Number : 327056-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171-173
Synonym 3-Carboxy-5-fluorobenzonitrile
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-cyano-5-fluorobenzoic-acid-98

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-cyano-5-fluorobenzoic-acid-98
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart