APO456796847 Apollo Scientific Ltd

4-(tert-Butoxy)-2,3,5,6-tetrafluorostyrene

  • MDL number MFCD04038273
  • Cas Number : 343305-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 52/ 0.04mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8237

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=8237
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart