APO456796851 Apollo Scientific Ltd

4-Fluoro-3-nitroanisole

  • MDL number MFCD11848442
  • Cas Number : 61324-93-4
  • Molecular Weight : 171.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52-53
Synonym 1-Fluoro-4-methoxy-2-nitrobenzene, 4-Fluoro-3-nitrophenyl methyl ether, 2-Fluoro-5-methoxynitrobenzene
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-3-nitroanisole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-3-nitroanisole
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart