APO456796866 Apollo Scientific Ltd

4-Bromo-2,3-difluorobenzeneboronic acid

  • MDL number MFCD02093065
  • Cas Number : 374790-99-5
  • Molecular Weight : 236.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225
Synonym 4-Borono-1-bromo-2,3-difluorobenzene
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-23-difluorobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/4-bromo-23-difluorobenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart