APO456796878 Apollo Scientific Ltd

4-(1,1,1,3,3,3-Hexafluoro-2-hydroxyprop-2-yl)benzoic acid

  • MDL number MFCD00236043
  • Cas Number : 16261-80-6
  • Molecular Weight : 288.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 121-123
Iupac Name 4-[2,2,2-trifluoro-1-hydroxy-1-(trifluoromethyl)ethyl]benzoic acid
Inchi Code 1S/C10H6F6O3/c11-9(12,13)8(19,10(14,15)16)6-3-1-5(2-4-6)7(17)18/h1-4,19H,(H,17,18)
Inchi Key DLJNNINHDYILFL-UHFFFAOYSA-N

Description

Synonym (4-Carboxyphenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol, 4-[Perfluoro(2-hydroxyprop-2-yl)]benzoic acid
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 121-123
Product Link https://store.apolloscientific.co.uk/product/4-111333-hexafluoro-2-hydroxyprop-2-ylbenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-111333-hexafluoro-2-hydroxyprop-2-ylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart