APO456796903 Apollo Scientific Ltd

2-Fluoro-alpha-methylbenzyl alcohol

  • MDL number MFCD00014401
  • Cas Number : 445-26-1
  • Molecular Weight : 140.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-fluorophenyl)ethanol
Inchi Code 1S/C8H9FO/c1-6(10)7-4-2-3-5-8(7)9/h2-6,10H,1H3
Inchi Key SXFYVXSOEBCFLV-UHFFFAOYSA-N

Description

Synonym 1-(2-Fluorophenyl)ethan-1-ol, 2-Fluorophenylmethylcarbinol
Flash Point 110/135mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-alpha-methylbenzyl-alcohol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-alpha-methylbenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart