APO456796985 Apollo Scientific Ltd

1H,1H,2H,2H,3H,3H-Perfluorohexan-1-ol

  • MDL number MFCD00077554
  • Cas Number : 679-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4,4,5,5,6,6,6-Heptafluorohexan-1-ol, 3-(Perfluoropropyl)propan-1-ol, 1,1,1,2,2,3,3-Heptafluoro-6-hydroxyhexane
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 140-141/745mm
Product Link https://store.apolloscientific.co.uk/product/1h1h2h2h3h3h-perfluorohexan-1-ol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1h1h2h2h3h3h-perfluorohexan-1-ol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart