APO456796992 Apollo Scientific Ltd

3,3,4,4,5,5,5-Heptafluoropentan-2-ol

  • MDL number MFCD00039619
  • Cas Number : 375-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Density 1.483
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 101-102
Product Link https://store.apolloscientific.co.uk/product/3344555-heptafluoropentan-2-ol-95

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3344555-heptafluoropentan-2-ol-95
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart