APO456797114 Apollo Scientific Ltd

1,4-Bis(pentafluorothio)perfluorobenzene

  • MDL number MFCD18071069
  • Cas Number : 1219501-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129-130
Synonym 1,4-Bis(pentafluorothio)-2,3,5,6-tetrafluorobenzene, 1,4-Bis(pentafluorosulphanyl)-2,3,5,6-tetrafluorobenzene
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48179

Safety Information

signalword Danger
pcodes P280;P210
MSDS http://www.apolloscientific.co.uk/display_item.php?id=48179
pictograms GHS02;GHS07
hstatements H228;H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart