APO456797157 Apollo Scientific Ltd

2-Fluoro-5-methoxyaniline

  • MDL number MFCD11226283
  • Cas Number : 62257-15-2
  • Molecular Weight : 141.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-fluoro-5-methoxyaniline
Inchi Code 1S/C7H8FNO/c1-10-5-2-3-6(8)7(9)4-5/h2-4H,9H2,1H3
Inchi Key RUTKJXMYXZFXPQ-UHFFFAOYSA-N

Description

Synonym 3-Amino-4-fluoroanisole, 6-Fluoro-m-anisidine
Storage temp. Store at 2-8
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-5-methoxyaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-5-methoxyaniline
pictograms GHS07
hstatements H302;H312;H332
Availability details for
Quantity
  • Availability
  • Add to Cart