APO456797215 Apollo Scientific Ltd

Manganese(III) fluoride

  • MDL number MFCD00016223
  • Cas Number : 7783-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Manganese trifluoride
Purity 99
Density 3.54
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3819

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=3819
pictograms GHS03;GHS06
hstatements H272;H301;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart