APO456797223 Apollo Scientific Ltd

2-Fluoro-5-(hydroxymethyl)benzeneboronic acid

  • MDL number MFCD06656276
  • Cas Number : 1072952-25-0
  • Molecular Weight : 169.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-76
Synonym 3-Borono-4-fluorobenzyl alcohol
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-5-hydroxymethylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P280;P302+P352;P321
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-5-hydroxymethylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart