APO456797231 Apollo Scientific Ltd

4-Fluoro-3-(hydroxymethyl)benzeneboronic acid

  • MDL number MFCD08235075
  • Cas Number : 481681-02-1
  • Molecular Weight : 169.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124-126
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-3-hydroxymethylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-3-hydroxymethylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart