APO456797284 Apollo Scientific Ltd

Methyl perfluorododecanoate

  • MDL number MFCD00236616
  • Cas Number : 56554-52-0
  • Molecular Weight : 628.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54-56
Purity 95
Density 1.66
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-perfluorododecanoate

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/methyl-perfluorododecanoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart