APO456797287 Apollo Scientific Ltd

Methyl 4,4,4-trifluoroacetoacetate

  • MDL number MFCD00041004
  • Cas Number : 83643-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 3-oxo-4,4,4-trifluorobutanoate, Methyl 3-oxo-4,4,4-trifluorobutyrate
Purity 95
Density 1.34
Flash Point 40
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 116-118
Product Link https://store.apolloscientific.co.uk/product/methyl-444-trifluoroacetoacetate-95

Safety Information

signalword Warning
pcodes P210;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-444-trifluoroacetoacetate-95
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart