APO456797392 Apollo Scientific Ltd

2-Amino-5-hydroxybenzotrifluoride

  • MDL number MFCD01862068
  • Cas Number : 445-04-5
  • Molecular Weight : 177.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-158
Synonym 4-Amino-3-(trifluoromethyl)phenol, 4-Hydroxy-2-(trifluoromethyl)aniline
Purity 98
Flash Point >110
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-5-hydroxybenzotrifluoride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-5-hydroxybenzotrifluoride
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart