APO456797418 Apollo Scientific Ltd

Methyl 5-chloro-3-fluoropyridine-2-carboxylate

  • MDL number MFCD14698145
  • Cas Number : 1200498-40-3
  • Molecular Weight : 189.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 5-chloro-3-fluoropicolinate, 5-Chloro-3-fluoro-2-(methoxycarbonyl)pyridine
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-5-chloro-3-fluoropyridine-2-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-5-chloro-3-fluoropyridine-2-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart