APO456797467 Apollo Scientific Ltd

1-Bromo-1H,1H,2H,2H-heptadecafluorodecane

  • MDL number MFCD04039296
  • Cas Number : 21652-57-3
  • Molecular Weight : 527.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 23.6-23.9
Synonym 2-(Perfluorooct-1-yl)ethyl bromide, 1-Bromo-2-(perfluorooct-1-yl)ethane, 10-Bromo-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluorodecane
Storage temp. Store at 2-8
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 84/10mm
Product Link https://store.apolloscientific.co.uk/product/1-bromo-1h1h2h2h-heptadecafluorodecane

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-bromo-1h1h2h2h-heptadecafluorodecane
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart