APO456797524 Apollo Scientific Ltd

2-Methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxaldehyde

  • MDL number MFCD12198128
  • Cas Number : 1034566-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Formyl-2-methyl-4-(trifluoromethyl)-1,3-thiazole
Purity 97
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=46620

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=46620
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart