APO456797527 Apollo Scientific Ltd

Methyl perfluoro(2-methyl-3-oxahexanoate)

  • MDL number MFCD00042358
  • Cas Number : 13140-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 2-(heptafluoropropoxy)-2,3,3,3-tetrafluoropropanoate, Methyl 2-(heptafluoropropoxy)-2,3,3,3-tetrafluoropropionate, Methyl undecafluoro-2-methyl-3-oxahexanoate
Purity 97
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 115-117
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3967

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=3967
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart