APO456797546 Apollo Scientific Ltd

1H,1H-Nonafluoropentan-1-ol

  • MDL number MFCD00236714
  • Cas Number : 355-28-2
  • Molecular Weight : 250.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (Perfluorobut-1-yl)methanol, 2,2,3,3,4,4,5,5,5-Nonafluoropentan-1-ol
Purity 97
Density 1.66
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 110-111
Product Link https://store.apolloscientific.co.uk/product/1h1h-nonafluoropentan-1-ol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1h1h-nonafluoropentan-1-ol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart