APO456797553 Apollo Scientific Ltd

Tris(perfluorohex-1-yl)amine

  • MDL number MFCD00042590
  • Cas Number : 432-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Tris(tridecafluorohex-1-yl)amine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 250-260
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4623

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=4623
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart