APO456797570 Apollo Scientific Ltd

2-Fluoro-4-(methylsulphanyl)benzeneboronic acid

  • MDL number MFCD09027247
  • Cas Number : 957060-84-3
  • Molecular Weight : 186.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 122-123
Iupac Name 2-fluoro-4-(methylsulfanyl)phenylboronic acid
Inchi Code 1S/C7H8BFO2S/c1-12-5-2-3-6(8(10)11)7(9)4-5/h2-4,10-11H,1H3
Inchi Key OUFPJVWVFXZKOX-UHFFFAOYSA-N

Description

Synonym 4-Borono-3-fluorothioanisole, 2-Fluoro-4-(methylthio)benzeneboronic acid
Purity 98%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 122-123
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-4-methylsulphanylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-4-methylsulphanylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart