APO456797581 Apollo Scientific Ltd

Sodium hexafluorotitanate(IV)

  • MDL number MFCD00014244
  • Cas Number : 17116-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/sodium-hexafluorotitanateiv-98

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/sodium-hexafluorotitanateiv-98
pictograms GHS07
hstatements EUH031;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart