APO456797621 Apollo Scientific Ltd

3-Amino-1-methyl-5-(trifluoromethyl)-1H-indazole

  • MDL number MFCD07777192
  • Cas Number : 5685-69-8
  • Molecular Weight : 215.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 140-142
Iupac Name 1-methyl-5-(trifluoromethyl)-1H-indazol-3-amine
Inchi Code 1S/C9H8F3N3/c1-15-7-3-2-5(9(10,11)12)4-6(7)8(13)14-15/h2-4H,1H3,(H2,13,14)
Inchi Key IRSFDHYOPSXICW-UHFFFAOYSA-N

Description

Synonym 1-Methyl-5-(trifluoromethyl)-1H-indazol-3-amine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 140-142
Product Link https://store.apolloscientific.co.uk/product/3-amino-1-methyl-5-trifluoromethyl-1h-indazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-1-methyl-5-trifluoromethyl-1h-indazole
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart