APO456797668 Apollo Scientific Ltd

1,1-Dichloro-2,2,2-trifluoroethyl difluoromethyl ether

  • MDL number MFCD01320775
  • Cas Number : 32778-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,1-Dichloro-1-(difluoromethoxy)-2,2,2-trifluoroethane, 2,2-Dichloro-1,1,1,4,4-pentafluoro-3-oxabutane
Purity 98
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 60-61
Product Link https://store.apolloscientific.co.uk/product/11-dichloro-222-trifluoroethyl-difluoromethyl-ether-98

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/11-dichloro-222-trifluoroethyl-difluoromethyl-ether-98
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart