APO456797747 Apollo Scientific Ltd

3,4-Difluoro-2-methylbenzeneboronic acid, pinacol ester

  • MDL number MFCD08669570
  • Cas Number : 1025707-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(3,4-Difluoro-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2,3-Difluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)toluene
Purity 95
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36210

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36210
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart