APO456797761 Apollo Scientific Ltd

2-(Trifluoromethyl)pyridine-3-boronic acid

  • MDL number MFCD09271791
  • Cas Number : 947533-39-3
  • Molecular Weight : 190.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(trifluoromethyl)-3-pyridinylboronic acid
Inchi Code 1S/C6H5BF3NO2/c8-6(9,10)5-4(7(12)13)2-1-3-11-5/h1-3,12-13H
Inchi Key RWKHOCBPUSCGAW-UHFFFAOYSA-N

Description

Synonym 3-Borono-2-(trifluoromethyl)pyridine
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-trifluoromethylpyridine-3-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-trifluoromethylpyridine-3-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart